The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008396.2 | NP_001008397.1 | 21 - 22 | 2741 | 64 |
NM_007086.3 | NP_009017.1 | 22 - 23 | 2834 | 64 | |
XM_006720012.2 | XP_006720075.1 | 22 - 23 | 2826 | 64 | |
XM_011536373.2 | XP_011534675.1 | 21 - 22 | 2633 | 64 | |
XM_017020940.1 | XP_016876429.1 | 21 - 22 | 2739 | 64 | |
XM_017020941.1 | XP_016876430.1 | 19 - 20 | 2437 | 64 | |
GenBank mRNA | AJ006266.1 | - | 21 - 22 | 2783 | 64 |
AK001538.1 | - | 10 - 11 | 1388 | 64 | |
AK001585.1 | - | 12 - 13 | 1757 | 64 | |
AK001620.1 | - | 5 - 6 | 561 | 64 | |
AK293005.1 | - | 22 - 23 | 2801 | 64 | |
BC063041.1 | - | 22 - 23 | 2810 | 64 |