The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008396.2 | NP_001008397.1 | 9 - 10 | 934 | 136 |
NM_007086.3 | NP_009017.1 | 10 - 11 | 1027 | 136 | |
XM_006720012.2 | XP_006720075.1 | 10 - 11 | 1025 | 136 | |
XM_017020940.1 | XP_016876429.1 | 9 - 10 | 938 | 136 | |
GenBank mRNA | AJ006266.1 | - | 9 - 10 | 976 | 136 |
AK293005.1 | - | 10 - 11 | 994 | 136 | |
BC000622.2 | - | 10 - 11 | 992 | 136 | |
BC005040.1 | - | 10 - 11 | 992 | 136 | |
BC043349.1 | - | 10 - 11 | 1015 | 136 | |
BC063041.1 | - | 10 - 11 | 1003 | 136 |