The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257096.1 | NP_001244025.1 | 1 - 2 | 340 | 157 |
NM_006192.4 | NP_006183.2 | 1 - 2 | 340 | 157 |