The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032816.4 | NP_116205.3 | 14 - 15 | 1726 | 89 |
XM_005259344.3 | XP_005259401.1 | 14 - 15 | 1649 | 89 | |
XM_011527425.2 | XP_011525727.1 | 11 - 12 | 1173 | 89 | |
XM_017027398.1 | XP_016882887.1 | 14 - 15 | 1721 | 89 | |
GenBank mRNA | AK027546.1 | - | 13 - 14 | 1427 | 89 |
AL832158.1 | - | 13 - 14 | 1549 | 89 | |
BC136328.1 | - | 14 - 15 | 1641 | 89 |