The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321643.1 | NP_001308572.1 | 17 - 18 | 2037 | 126 |
NM_004850.4 | NP_004841.2 | 17 - 18 | 2493 | 126 | |
XM_005246190.3 | XP_005246247.1 | 17 - 18 | 2527 | 126 | |
XM_011510417.2 | XP_011508719.1 | 17 - 18 | 1992 | 126 | |
XM_017005378.1 | XP_016860867.1 | 17 - 18 | 2524 | 126 | |
XM_017005379.1 | XP_016860868.1 | 17 - 18 | 1922 | 126 | |
GenBank mRNA | AB014519.1 | - | 17 - 18 | 2498 | 126 |
AK299296.1 | - | 3 - 4 | 500 | 126 | |
AK310358.1 | - | 13 - 14 | 1788 | 126 | |
BC111801.1 | - | 4 - 5 | 600 | 126 | |
D87931.1 | - | 17 - 18 | 2044 | 126 |