Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ROCK2
Entrez Gene ID
9475
Gene Name
Rho associated coiled-coil containing protein kinase 2
Gene Aliases
ROCK-II
Chromosome Location
Chr.2: 11179759 - 11345407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321643.1 NP_001308572.1 24 - 25 2993 107
NM_004850.4 NP_004841.2 24 - 25 3449 107
XM_005246190.3 XP_005246247.1 24 - 25 3483 107
XM_011510417.2 XP_011508719.1 24 - 25 2948 107
XM_017005378.1 XP_016860867.1 24 - 25 3480 107
XM_017005379.1 XP_016860868.1 24 - 25 2878 107
GenBank mRNA AB014519.1 - 24 - 25 3454 107
AK299296.1 - 10 - 11 1456 107
AK310358.1 - 20 - 21 2744 107
BC111801.1 - 11 - 12 1556 107
D87931.1 - 24 - 25 3000 107

View Full Product Details