The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321643.1 | NP_001308572.1 | 26 - 27 | 3275 | 89 |
NM_004850.4 | NP_004841.2 | 26 - 27 | 3731 | 89 | |
XM_005246190.3 | XP_005246247.1 | 26 - 27 | 3765 | 89 | |
XM_011510417.2 | XP_011508719.1 | 26 - 27 | 3230 | 89 | |
XM_017005378.1 | XP_016860867.1 | 26 - 27 | 3762 | 89 | |
XM_017005379.1 | XP_016860868.1 | 26 - 27 | 3160 | 89 | |
GenBank mRNA | AB014519.1 | - | 26 - 27 | 3736 | 89 |
AK299296.1 | - | 12 - 13 | 1738 | 89 | |
AK310358.1 | - | 22 - 23 | 3026 | 89 | |
BC111801.1 | - | 13 - 14 | 1838 | 89 | |
D87931.1 | - | 26 - 27 | 3282 | 89 |