The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000329.2 | NP_000320.1 | 5 - 6 | 551 | 98 |
XM_017002027.1 | XP_016857516.1 | 4 - 5 | 763 | 98 | |
GenBank mRNA | AK289925.1 | - | 5 - 6 | 542 | 98 |
BC075035.2 | - | 5 - 6 | 543 | 98 | |
BC075036.2 | - | 5 - 6 | 546 | 98 | |
U18991.1 | - | 5 - 6 | 551 | 98 |