The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000255.3 | NP_000246.2 | 5 - 6 | 1346 | 105 |
XM_005249143.3 | XP_005249200.1 | 5 - 6 | 1251 | 105 | |
GenBank mRNA | AK292568.1 | - | 5 - 6 | 1322 | 105 |
AK312611.1 | - | 5 - 6 | 1321 | 105 | |
BC016282.1 | - | 5 - 6 | 1209 | 105 | |
BT007434.1 | - | 4 - 5 | 1081 | 105 | |
BX647789.1 | - | 5 - 6 | 1171 | 105 | |
KF030882.1 | - | 4 - 5 | 1081 | 105 | |
KU178201.1 | - | 4 - 5 | 1081 | 105 | |
KU178202.1 | - | 4 - 5 | 1081 | 105 | |
M65131.1 | - | 5 - 6 | 1157 | 105 |