The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146015.1 | NP_001139487.1 | 4 - 5 | 709 | 73 |
NM_014750.4 | NP_055565.3 | 4 - 5 | 709 | 73 | |
XM_017021840.1 | XP_016877329.1 | 4 - 5 | 601 | 73 | |
GenBank mRNA | AB076695.1 | - | 4 - 5 | 709 | 73 |
AK291847.1 | - | 4 - 5 | 586 | 73 | |
AK299338.1 | - | 4 - 5 | 586 | 73 | |
BC010658.1 | - | 4 - 5 | 601 | 73 | |
BC016276.2 | - | 3 - 4 | 495 | 73 | |
BT007344.1 | - | 2 - 3 | 249 | 73 | |
BX248255.1 | - | 4 - 5 | 575 | 73 | |
D13633.3 | - | 4 - 5 | 582 | 73 |