The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016052.3 | NP_057136.2 | 1 - 2 | 169 | 86 |
XM_011509597.2 | XP_011507899.1 | 1 - 2 | 186 | 86 | |
GenBank mRNA | AF151873.1 | - | 1 - 2 | 104 | 86 |
AK290066.1 | - | 1 - 2 | 151 | 86 | |
BC003668.1 | - | 1 - 2 | 147 | 86 | |
BC020641.1 | - | 1 - 2 | 146 | 86 | |
BF242786.1 | - | 1 - 2 | 170 | 86 |