Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
Chromosome Location
Chr.11: 72223861 - 72239147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001567.3 NP_001558.3 6 - 7 899 54
XM_005273978.3 XP_005274035.1 7 - 8 1276 54
XM_005273979.4 XP_005274036.1 7 - 8 1123 54
XM_011544999.2 XP_011543301.1 7 - 8 1057 54
XM_011545000.2 XP_011543302.1 7 - 8 1123 54
XM_017017673.1 XP_016873162.1 8 - 9 1214 54
GenBank mRNA BC140853.1 - 6 - 7 899 54
L36818.1 - 5 - 6 876 54
Y14385.1 - 6 - 7 963 54

View Full Product Details