The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006244.3 | NP_006235.1 | 12 - 13 | 1864 | 71 |
XM_011545132.2 | XP_011543434.1 | 13 - 14 | 1297 | 71 | |
XM_011545133.1 | XP_011543435.1 | 14 - 15 | 1417 | 71 | |
XM_011545134.2 | XP_011543436.1 | 12 - 13 | 2703 | 71 | |
GenBank mRNA | BC045619.1 | - | 12 - 13 | 1858 | 71 |
L42374.1 | - | 12 - 13 | 1567 | 71 |