The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098722.1 | NP_001092192.1 | 1 - 2 | 334 | 61 |
GenBank mRNA | AK311781.1 | - | 1 - 2 | 123 | 61 |
CX784976.1 | - | 1 - 2 | 283 | 61 | |
DA122711.1 | - | 1 - 2 | 114 | 61 |