Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRC5
Entrez Gene ID
84166
Gene Name
NLR family CARD domain containing 5
Gene Aliases
CLR16.1, NOD27, NOD4
Chromosome Location
Chr.16: 56989547 - 57083524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032206.4 NP_115582.4 19 - 20 3336 79
XM_005256193.2 XP_005256250.1 20 - 21 3397 79
XM_005256194.3 XP_005256251.1 20 - 21 3608 79
XM_005256195.2 XP_005256252.1 20 - 21 3397 79
XM_005256196.2 XP_005256253.1 19 - 20 3313 79
XM_005256197.2 XP_005256254.1 20 - 21 3397 79
XM_005256199.3 XP_005256256.1 20 - 21 3397 79
XM_006721297.2 XP_006721360.1 20 - 21 3385 79
XM_006721300.2 XP_006721363.1 20 - 21 3397 79
XM_011523373.2 XP_011521675.1 20 - 21 3544 79
XM_011523375.2 XP_011521677.1 20 - 21 3397 79
XM_011523376.2 XP_011521678.1 15 - 16 1223 79
XM_017023766.1 XP_016879255.1 20 - 21 3397 79
XM_017023767.1 XP_016879256.1 20 - 21 3385 79
XM_017023768.1 XP_016879257.1 20 - 21 3397 79
XM_017023769.1 XP_016879258.1 20 - 21 3385 79
XM_017023770.1 XP_016879259.1 20 - 21 3397 79
XM_017023771.1 XP_016879260.1 20 - 21 3397 79
GenBank mRNA AF389420.1 - 18 - 19 3209 79
AK025362.1 - 15 - 16 2468 79
AK074133.1 - 8 - 9 662 79
AK074182.1 - 20 - 21 3832 79
AK090439.1 - 15 - 16 2391 79
BC050527.1 - 4 - 5 301 79
BC063566.1 - 2 - 3 122 79
EF452236.1 - 18 - 19 3209 79

View Full Product Details