The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032206.4 | NP_115582.4 | 38 - 39 | 4875 | 80 |
XM_005256193.2 | XP_005256250.1 | 39 - 40 | 4936 | 80 | |
XM_005256194.3 | XP_005256251.1 | 39 - 40 | 5147 | 80 | |
XM_005256195.2 | XP_005256252.1 | 39 - 40 | 4933 | 80 | |
XM_005256196.2 | XP_005256253.1 | 38 - 39 | 4852 | 80 | |
XM_005256197.2 | XP_005256254.1 | 38 - 39 | 4849 | 80 | |
XM_006721297.2 | XP_006721360.1 | 39 - 40 | 4924 | 80 | |
XM_006721298.2 | XP_006721361.1 | 38 - 39 | 4846 | 80 | |
XM_011523373.2 | XP_011521675.1 | 39 - 40 | 5083 | 80 | |
XM_011523376.2 | XP_011521678.1 | 34 - 35 | 2762 | 80 | |
XM_017023767.1 | XP_016879256.1 | 38 - 39 | 4837 | 80 | |
GenBank mRNA | AF389420.1 | - | 37 - 38 | 4748 | 80 |
AK027414.1 | - | 18 - 19 | 1465 | 80 | |
AK027416.1 | - | 18 - 19 | 1464 | 80 | |
AK074133.1 | - | 27 - 28 | 2265 | 80 | |
AK090439.1 | - | 34 - 35 | 3988 | 80 | |
BC050527.1 | - | 23 - 24 | 1768 | 80 | |
BC063566.1 | - | 21 - 22 | 1661 | 80 | |
EF452236.1 | - | 37 - 38 | 4748 | 80 |