Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC68
Entrez Gene ID
80323
Gene Name
coiled-coil domain containing 68
Gene Aliases
SE57-1
Chromosome Location
Chr.18: 54901509 - 54959539 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143829.1 NP_001137301.1 6 - 7 649 153
NM_025214.2 NP_079490.1 6 - 7 752 153
XM_006722552.3 XP_006722615.1 6 - 7 701 153
XM_011526198.2 XP_011524500.1 6 - 7 1761 153
XM_011526199.2 XP_011524501.1 6 - 7 660 153
XM_011526200.2 XP_011524502.1 6 - 7 813 153
XM_011526201.2 XP_011524503.1 6 - 7 1720 153
XM_011526202.1 XP_011524504.1 6 - 7 752 153
XM_011526203.2 XP_011524505.1 5 - 6 587 153
XM_011526204.2 XP_011524506.1 6 - 7 752 153
XM_017026011.1 XP_016881500.1 5 - 6 753 153
GenBank mRNA AF273051.1 - 6 - 7 650 153
AK058097.1 - 6 - 7 649 153
AK313793.1 - 4 - 5 495 153
BC029508.1 - 6 - 7 706 153

View Full Product Details