The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011537237.2 | XP_011535539.1 | 1 - 2 | 919 | 120 |
GenBank mRNA | AB593096.1 | - | 1 - 2 | 118 | 120 |
AB593097.1 | - | 1 - 2 | 351 | 120 | |
BC132877.1 | - | 1 - 2 | 286 | 120 | |
BC136577.1 | - | 1 - 2 | 286 | 120 |