Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HHIPL1
Entrez Gene ID
84439
Gene Name
HHIP like 1
Gene Aliases
KIAA1822, UNQ9245
Chromosome Location
Chr.14: 99604476 - 99680569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006720277.3 XP_006720340.1 5 - 6 1489 79
XM_011537236.2 XP_011535538.1 5 - 6 1607 79
XM_011537237.2 XP_011535539.1 4 - 5 2042 79
XM_017021707.1 XP_016877196.1 4 - 5 1441 79
XM_017021708.1 XP_016877197.1 5 - 6 1607 79
GenBank mRNA AB058725.1 - 3 - 4 631 79
AB593096.1 - 4 - 5 1241 79
AB593097.1 - 4 - 5 1474 79
AY358173.1 - 4 - 5 1441 79
BC132877.1 - 4 - 5 1409 79
BC136577.1 - 4 - 5 1409 79

View Full Product Details