The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006720277.3 | XP_006720340.1 | 7 - 8 | 1765 | 87 |
XM_011537236.2 | XP_011535538.1 | 7 - 8 | 1883 | 87 | |
XM_011537237.2 | XP_011535539.1 | 6 - 7 | 2318 | 87 | |
XM_017021707.1 | XP_016877196.1 | 6 - 7 | 1717 | 87 | |
XM_017021708.1 | XP_016877197.1 | 7 - 8 | 1883 | 87 | |
GenBank mRNA | AB058725.1 | - | 5 - 6 | 907 | 87 |
AB593096.1 | - | 6 - 7 | 1517 | 87 | |
AB593097.1 | - | 6 - 7 | 1750 | 87 | |
AY358173.1 | - | 6 - 7 | 1717 | 87 | |
BC132877.1 | - | 6 - 7 | 1685 | 87 | |
BC136577.1 | - | 6 - 7 | 1685 | 87 |