The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005691.3 | NP_005682.2 | 22 - 23 | 2792 | 91 |
NM_020297.3 | NP_064693.2 | 22 - 23 | 2792 | 91 | |
XM_005253284.3 | XP_005253341.1 | 24 - 25 | 3578 | 91 | |
XM_005253286.3 | XP_005253343.1 | 25 - 26 | 3638 | 91 | |
XM_005253287.4 | XP_005253344.1 | 24 - 25 | 3578 | 91 | |
XM_005253288.3 | XP_005253345.1 | 23 - 24 | 3462 | 91 | |
XM_005253289.3 | XP_005253346.1 | 23 - 24 | 3539 | 91 | |
XM_005253290.3 | XP_005253347.1 | 22 - 23 | 3437 | 91 | |
XM_006719025.3 | XP_006719088.1 | 23 - 24 | 3539 | 91 | |
XM_011520545.2 | XP_011518847.1 | 24 - 25 | 3634 | 91 | |
GenBank mRNA | CA389699.1 | - | 1 - 2 | 89 | 91 |