Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTL8
Entrez Gene ID
81569
Gene Name
actin like 8
Gene Aliases
CT57
Chromosome Location
Chr.1: 17755313 - 17827063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030812.2 NP_110439.2 1 - 2 197 90
GenBank mRNA AK057339.1 - 1 - 2 197 90
BC028909.1 - 1 - 2 161 90
BI826094.1 - 1 - 2 149 90
U20582.1 - 1 - 2 118 90

View Full Product Details