Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNIH3
Entrez Gene ID
149111
Gene Name
cornichon family AMPA receptor auxiliary protein 3
Gene Aliases
CNIH-3
Chromosome Location
Chr.1: 224434640 - 224740549 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322302.1 NP_001309231.1 6 - 7 735 84
NM_001322303.1 NP_001309232.1 5 - 6 669 84
NM_001322304.1 NP_001309233.1 6 - 7 813 84
NM_001322305.1 NP_001309234.1 4 - 5 1107 84
NM_152495.1 NP_689708.1 5 - 6 1155 84
NR_136287.1 4 - 5 1042 84
NR_136288.1 6 - 7 665 84
NR_136289.1 7 - 8 801 84
NR_136290.1 6 - 7 735 84
NR_136291.1 7 - 8 871 84
NR_136292.1 6 - 7 805 84
NR_136293.1 9 - 10 1041 84
NR_136294.1 8 - 9 975 84
NR_136295.1 7 - 8 846 84
NR_136296.1 7 - 8 871 84
NR_136297.1 6 - 7 735 84
GenBank mRNA AF070524.1 - 4 - 5 361 84
AK096312.1 - 5 - 6 1155 84
BC022780.1 - 5 - 6 643 84

View Full Product Details