Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR10H1
Entrez Gene ID
26539
Gene Name
olfactory receptor family 10 subfamily H member 1
Gene Aliases
-
Chromosome Location
Chr.19: 15806957 - 15808759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013940.2 NP_039228.1 1 - 1 991 69
XM_011527911.1 XP_011526213.1 2 - 2 948 69
GenBank mRNA BC130360.1 - 1 - 1 991 69
BC132969.1 - 1 - 1 991 69

View Full Product Details