Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM17
Entrez Gene ID
200728
Gene Name
transmembrane protein 17
Gene Aliases
-
Chromosome Location
Chr.2: 62452778 - 62535645 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198276.2 NP_938017.2 3 - 4 660 123
XM_017003575.1 XP_016859064.1 4 - 5 571 123
GenBank mRNA BC047439.1 - 3 - 4 532 123
DR760587.1 - 3 - 4 697 123

View Full Product Details