The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138295.3 | NP_612152.1 | 51 - 52 | 7722 | 84 |
XM_017011798.1 | XP_016867287.1 | 52 - 53 | 14127 | 84 | |
GenBank mRNA | AB061683.1 | - | 51 - 52 | 7688 | 84 |