The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183242.3 | NP_899065.2 | 5 - 6 | 1018 | 135 |
GenBank mRNA | BC013922.1 | - | 5 - 6 | 991 | 135 |