The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002410.4 | NP_002401.1 | 15 - 16 | 2164 | 76 |
XM_005263668.4 | XP_005263725.1 | 16 - 17 | 2529 | 76 | |
XM_005263669.4 | XP_005263726.1 | 16 - 17 | 2526 | 76 | |
XM_005263670.3 | XP_005263727.1 | 16 - 17 | 2182 | 76 | |
XM_006712534.3 | XP_006712597.1 | 17 - 18 | 2378 | 76 | |
XM_011511199.2 | XP_011509501.1 | 17 - 18 | 2813 | 76 | |
XM_011511200.1 | XP_011509502.1 | 16 - 17 | 2178 | 76 | |
XM_011511201.2 | XP_011509503.1 | 16 - 17 | 2282 | 76 | |
XM_011511202.1 | XP_011509504.1 | 16 - 17 | 2179 | 76 | |
XM_017004147.1 | XP_016859636.1 | 17 - 18 | 2653 | 76 | |
XM_017004148.1 | XP_016859637.1 | 16 - 17 | 2490 | 76 | |
GenBank mRNA | AF113921.1 | - | 15 - 16 | 2052 | 76 |
D17716.1 | - | 15 - 16 | 2164 | 76 |