Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGAT5
Entrez Gene ID
4249
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase
Gene Aliases
GNT-V, GNT-VA
Chromosome Location
Chr.2: 134119922 - 134454621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002410.4 NP_002401.1 2 - 3 553 83
XM_005263668.4 XP_005263725.1 3 - 4 918 83
XM_005263669.4 XP_005263726.1 3 - 4 915 83
XM_005263670.3 XP_005263727.1 3 - 4 571 83
XM_006712534.3 XP_006712597.1 4 - 5 767 83
XM_011511199.2 XP_011509501.1 4 - 5 1202 83
XM_011511200.1 XP_011509502.1 3 - 4 567 83
XM_011511201.2 XP_011509503.1 3 - 4 671 83
XM_011511202.1 XP_011509504.1 3 - 4 568 83
XM_011511204.2 XP_011509506.1 4 - 5 770 83
XM_017004147.1 XP_016859636.1 4 - 5 1042 83
XM_017004148.1 XP_016859637.1 3 - 4 879 83
XM_017004149.1 XP_016859638.1 3 - 4 920 83
GenBank mRNA AF113921.1 - 2 - 3 441 83
D17716.1 - 2 - 3 553 83

View Full Product Details