The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000720.3 | NP_000711.1 | 2 - 3 | 500 | 98 |
NM_001128839.2 | NP_001122311.1 | 2 - 3 | 500 | 98 | |
NM_001128840.2 | NP_001122312.1 | 2 - 3 | 500 | 98 | |
XM_005265448.3 | XP_005265505.1 | 2 - 3 | 382 | 98 | |
XM_011534094.2 | XP_011532396.1 | 2 - 3 | 508 | 98 | |
XM_011534096.2 | XP_011532398.1 | 2 - 3 | 508 | 98 | |
XM_011534100.2 | XP_011532402.1 | 2 - 3 | 508 | 98 | |
XM_017007137.1 | XP_016862626.1 | 2 - 3 | 508 | 98 | |
XM_017007138.1 | XP_016862627.1 | 2 - 3 | 508 | 98 | |
XM_017007139.1 | XP_016862628.1 | 2 - 3 | 508 | 98 | |
XM_017007140.1 | XP_016862629.1 | 2 - 3 | 508 | 98 | |
XM_017007141.1 | XP_016862630.1 | 2 - 3 | 508 | 98 | |
XM_017007142.1 | XP_016862631.1 | 2 - 3 | 508 | 98 | |
XM_017007143.1 | XP_016862632.1 | 2 - 3 | 508 | 98 | |
XM_017007144.1 | XP_016862633.1 | 2 - 3 | 508 | 98 | |
XM_017007145.1 | XP_016862634.1 | 2 - 3 | 508 | 98 | |
GenBank mRNA | EU363339.1 | - | 2 - 3 | 382 | 98 |
M76558.1 | - | 2 - 3 | 892 | 98 | |
M83566.1 | - | 2 - 3 | 500 | 98 |