Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1D
Entrez Gene ID
776
Gene Name
calcium voltage-gated channel subunit alpha1 D
Gene Aliases
CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
Chromosome Location
Chr.3: 53495049 - 53813151 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000720.3 NP_000711.1 47 - 48 6046 57
NM_001128839.2 NP_001122311.1 44 - 45 5914 57
NM_001128840.2 NP_001122312.1 46 - 47 5986 57
XM_005265448.3 XP_005265505.1 45 - 46 5823 57
XM_011534094.2 XP_011532396.1 48 - 49 6138 57
XM_011534096.2 XP_011532398.1 45 - 46 5949 57
XM_011534097.2 XP_011532399.1 46 - 47 6605 57
XM_011534099.2 XP_011532401.1 44 - 45 5588 57
XM_011534100.2 XP_011532402.1 46 - 47 6033 57
XM_017007137.1 XP_016862626.1 48 - 49 6138 57
XM_017007138.1 XP_016862627.1 48 - 49 6135 57
XM_017007139.1 XP_016862628.1 47 - 48 6111 57
XM_017007140.1 XP_016862629.1 47 - 48 6078 57
XM_017007141.1 XP_016862630.1 47 - 48 6078 57
XM_017007142.1 XP_016862631.1 47 - 48 6054 57
XM_017007143.1 XP_016862632.1 47 - 48 6054 57
XM_017007144.1 XP_016862633.1 47 - 48 6054 57
XM_017007145.1 XP_016862634.1 46 - 47 6009 57
GenBank mRNA AB209171.1 - 41 - 42 4947 57
EU363339.1 - 44 - 45 5796 57
M76558.1 - 46 - 47 6378 57
M83566.1 - 47 - 48 6046 57

View Full Product Details