The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012414.3 | NP_036546.2 | 17 - 18 | 1945 | 151 |
GenBank mRNA | AB020646.2 | - | 17 - 18 | 1781 | 151 |
AF004828.1 | - | 17 - 18 | 1823 | 151 | |
AF255648.1 | - | 17 - 18 | 1854 | 151 | |
AK291234.1 | - | 17 - 18 | 1896 | 151 | |
BC131573.1 | - | 17 - 18 | 1828 | 151 | |
BC146760.1 | - | 17 - 18 | 1781 | 151 |