The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000882.3 | NP_000873.2 | 1 - 2 | 340 | 52 |
GenBank mRNA | AF101062.1 | - | 1 - 2 | 119 | 52 |
AF180562.1 | - | 1 - 2 | 178 | 52 | |
BC104982.1 | - | 1 - 2 | 272 | 52 | |
BC104984.1 | - | 1 - 2 | 272 | 52 | |
BG702253.1 | - | 1 - 2 | 433 | 52 | |
M65271.1 | - | 1 - 2 | 186 | 52 |