The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243088.1 | NP_001230017.1 | 3 - 4 | 934 | 86 |
NM_001243089.1 | NP_001230018.1 | 3 - 4 | 934 | 86 | |
NM_021953.3 | NP_068772.2 | 3 - 4 | 937 | 86 | |
NM_202002.2 | NP_973731.1 | 3 - 4 | 937 | 86 | |
NM_202003.2 | NP_973732.1 | 3 - 4 | 937 | 86 | |
XM_005253676.3 | XP_005253733.1 | 3 - 4 | 937 | 86 | |
XM_011520930.2 | XP_011519232.1 | 3 - 4 | 934 | 86 | |
XM_011520931.2 | XP_011519233.1 | 3 - 4 | 937 | 86 | |
XM_011520932.1 | XP_011519234.1 | 3 - 4 | 937 | 86 | |
XM_011520933.1 | XP_011519235.1 | 3 - 4 | 937 | 86 | |
XM_011520934.2 | XP_011519236.1 | 3 - 4 | 937 | 86 | |
XM_011520935.1 | XP_011519237.1 | 3 - 4 | 937 | 86 | |
GenBank mRNA | AK291206.1 | - | 3 - 4 | 837 | 86 |
AK313845.1 | - | 3 - 4 | 780 | 86 | |
AY542306.1 | - | 2 - 3 | 705 | 86 | |
BC006192.2 | - | 3 - 4 | 822 | 86 | |
BC006529.2 | - | 3 - 4 | 822 | 86 | |
BC012863.1 | - | 2 - 3 | 673 | 86 | |
BT006986.1 | - | 2 - 3 | 654 | 86 | |
U74612.1 | - | 3 - 4 | 768 | 86 | |
U74613.1 | - | 3 - 4 | 761 | 86 | |
U83113.1 | - | 3 - 4 | 723 | 86 |