The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040458.1 | NP_001035548.1 | 12 - 13 | 2107 | 130 |
NM_001198541.1 | NP_001185470.1 | 12 - 13 | 1865 | 130 | |
NM_016442.3 | NP_057526.3 | 12 - 13 | 2107 | 130 | |
XM_005272015.4 | XP_005272072.1 | 12 - 13 | 2058 | 130 | |
XM_005272016.4 | XP_005272073.1 | 12 - 13 | 2018 | 130 | |
XM_011543480.2 | XP_011541782.1 | 16 - 17 | 4327 | 130 | |
XM_011543481.2 | XP_011541783.1 | 16 - 17 | 4324 | 130 | |
XM_011543484.2 | XP_011541786.1 | 16 - 17 | 4323 | 130 | |
XM_011543485.2 | XP_011541787.1 | 15 - 16 | 4143 | 130 | |
XM_011543486.2 | XP_011541788.1 | 16 - 17 | 4327 | 130 | |
XM_017009580.1 | XP_016865069.1 | 17 - 18 | 4383 | 130 | |
XM_017009581.1 | XP_016865070.1 | 15 - 16 | 4169 | 130 | |
XM_017009582.1 | XP_016865071.1 | 12 - 13 | 1842 | 130 | |
XM_017009583.1 | XP_016865072.1 | 12 - 13 | 2000 | 130 | |
GenBank mRNA | AB011097.2 | - | 11 - 12 | 1781 | 130 |
AF106037.1 | - | 12 - 13 | 1822 | 130 | |
AF183569.1 | - | 12 - 13 | 1827 | 130 | |
AF222340.1 | - | 12 - 13 | 1847 | 130 | |
AK291636.1 | - | 12 - 13 | 1857 | 130 | |
AK310535.1 | - | 11 - 12 | 1282 | 130 | |
AY028806.1 | - | 12 - 13 | 1857 | 130 | |
AY028807.1 | - | 12 - 13 | 1857 | 130 | |
AY358590.1 | - | 12 - 13 | 1845 | 130 | |
BC030775.1 | - | 12 - 13 | 1859 | 130 | |
JX962701.1 | - | 11 - 12 | 1760 | 130 | |
JX962702.1 | - | 11 - 12 | 1760 | 130 | |
KM357870.1 | - | 11 - 12 | 1760 | 130 | |
KM357871.1 | - | 11 - 12 | 1760 | 130 | |
KM357872.1 | - | 11 - 12 | 1760 | 130 | |
KM357873.1 | - | 11 - 12 | 1760 | 130 | |
KM357874.1 | - | 11 - 12 | 1760 | 130 | |
KM357875.1 | - | 11 - 12 | 1760 | 130 | |
KM357876.1 | - | 11 - 12 | 1760 | 130 | |
KM357877.1 | - | 11 - 12 | 1760 | 130 | |
KM357878.1 | - | 11 - 12 | 1760 | 130 | |
KM357879.1 | - | 11 - 12 | 1760 | 130 | |
KM357880.1 | - | 11 - 12 | 1760 | 130 | |
KM357881.1 | - | 11 - 12 | 1760 | 130 | |
KM357882.1 | - | 11 - 12 | 1760 | 130 | |
KM357883.1 | - | 11 - 12 | 1760 | 130 | |
KM357884.1 | - | 11 - 12 | 1760 | 130 | |
KM357885.1 | - | 11 - 12 | 1760 | 130 | |
KM357886.1 | - | 11 - 12 | 1760 | 130 | |
KM357887.1 | - | 11 - 12 | 1760 | 130 | |
KM357888.1 | - | 11 - 12 | 1760 | 130 | |
KM357889.1 | - | 11 - 12 | 1760 | 130 | |
KM357890.1 | - | 11 - 12 | 1760 | 130 | |
KM357891.1 | - | 11 - 12 | 1760 | 130 |