The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130969.1 | NP_001124441.1 | 13 - 14 | 1546 | 78 |
NM_001130970.1 | NP_001124442.1 | 12 - 13 | 1477 | 78 | |
NM_001130971.1 | NP_001124443.1 | 11 - 12 | 1471 | 78 | |
NM_001178064.1 | NP_001171535.1 | 12 - 13 | 1456 | 78 | |
NM_015537.4 | NP_056352.3 | 12 - 13 | 1540 | 78 | |
XM_005266061.4 | XP_005266118.1 | 11 - 12 | 1473 | 78 | |
XM_005266062.4 | XP_005266119.1 | 10 - 11 | 1404 | 78 | |
XM_011518496.2 | XP_011516798.1 | 11 - 12 | 1410 | 78 | |
XM_011518497.2 | XP_011516799.1 | 8 - 9 | 660 | 78 | |
XM_017014597.1 | XP_016870086.1 | 7 - 8 | 929 | 78 | |
GenBank mRNA | AJ420418.1 | - | 5 - 6 | 321 | 78 |
AK027474.1 | - | 10 - 11 | 811 | 78 | |
AK074602.1 | - | 12 - 13 | 1508 | 78 | |
AK292030.1 | - | 8 - 9 | 612 | 78 | |
AK294320.1 | - | 8 - 9 | 589 | 78 | |
AK303703.1 | - | 10 - 11 | 668 | 78 | |
AK303976.1 | - | 8 - 9 | 707 | 78 | |
AK316046.1 | - | 10 - 11 | 862 | 78 | |
AL117660.2 | - | 8 - 9 | 568 | 78 | |
AL389944.1 | - | 8 - 9 | 594 | 78 | |
AL389945.1 | - | 6 - 7 | 1440 | 78 | |
AL389946.1 | - | 4 - 5 | 234 | 78 | |
AY255128.1 | - | 13 - 14 | 1546 | 78 | |
AY255129.1 | - | 12 - 13 | 1540 | 78 | |
AY255130.1 | - | 12 - 13 | 1477 | 78 | |
AY255131.1 | - | 12 - 13 | 1456 | 78 | |
AY255132.1 | - | 12 - 13 | 1782 | 78 | |
BC004318.1 | - | 4 - 5 | 214 | 78 | |
BC016862.2 | - | 8 - 9 | 587 | 78 | |
BC072412.1 | - | 12 - 13 | 1525 | 78 | |
BC110498.2 | - | 11 - 12 | 1338 | 78 |