Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41L4B
Entrez Gene ID
54566
Gene Name
erythrocyte membrane protein band 4.1 like 4B
Gene Aliases
CG1, EHM2
Chromosome Location
Chr.9: 109171974 - 109320964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018424.3 NP_060894.2 11 - 12 1688 97
NM_019114.4 NP_061987.3 11 - 12 1688 97
XM_011518790.1 XP_011517092.1 11 - 12 1185 97
XM_011518791.2 XP_011517093.1 11 - 12 1170 97
XM_011518792.2 XP_011517094.1 11 - 12 1170 97
XM_011518794.2 XP_011517096.1 11 - 12 1170 97
XM_017014813.1 XP_016870302.1 11 - 12 1170 97
XM_017014814.1 XP_016870303.1 11 - 12 1170 97
XM_017014815.1 XP_016870304.1 11 - 12 1170 97
GenBank mRNA AB032179.3 - 11 - 12 1688 97
AB209187.1 - 3 - 4 280 97
AF153416.1 - 11 - 12 1460 97
AF153418.1 - 11 - 12 1460 97

View Full Product Details