Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
Chromosome Location
Chr.1: 151510859 - 151538692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020770.2 NP_065821.1 18 - 19 3335 76
XM_005245365.4 XP_005245422.1 18 - 19 3347 76
GenBank mRNA AB037740.1 - 18 - 19 3308 76
AF263462.1 - 18 - 19 3342 76
AK290007.1 - 18 - 19 3308 76
BC146657.1 - 17 - 18 3613 76
BC150604.1 - 17 - 18 3214 76
BC152445.1 - 18 - 19 3308 76
BX647343.1 - 3 - 4 1189 76

View Full Product Details