The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019029.2 | NP_061902.2 | 7 - 8 | 821 | 127 |
NM_031311.3 | NP_112601.3 | 7 - 8 | 733 | 127 | |
XM_011515437.1 | XP_011513739.1 | 7 - 8 | 900 | 127 | |
XM_017012366.1 | XP_016867855.1 | 7 - 8 | 733 | 127 | |
GenBank mRNA | AF106704.2 | - | 7 - 8 | 653 | 127 |
AF217508.1 | - | 6 - 7 | 620 | 127 | |
AF282617.1 | - | 7 - 8 | 683 | 127 | |
AK075433.1 | - | 7 - 8 | 734 | 127 | |
AK124472.1 | - | 11 - 12 | 1261 | 127 | |
AK225545.1 | - | 7 - 8 | 684 | 127 | |
AY358549.1 | - | 7 - 8 | 710 | 127 | |
BC016838.2 | - | 7 - 8 | 781 | 127 |