The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022459.4 | NP_071904.4 | 15 - 16 | 2185 | 92 |
XM_005266502.1 | XP_005266559.1 | 16 - 17 | 2340 | 92 | |
XM_005266503.3 | XP_005266560.1 | 15 - 16 | 2185 | 92 | |
XM_005266504.3 | XP_005266561.1 | 9 - 10 | 1712 | 92 | |
XM_011535197.1 | XP_011533499.1 | 16 - 17 | 2349 | 92 | |
XM_011535198.1 | XP_011533500.1 | 14 - 15 | 2027 | 92 | |
XM_011535199.1 | XP_011533501.1 | 13 - 14 | 1990 | 92 | |
XM_011535200.2 | XP_011533502.1 | 13 - 14 | 3440 | 92 | |
XM_017020704.1 | XP_016876193.1 | 11 - 12 | 1981 | 92 | |
XM_017020705.1 | XP_016876194.1 | 12 - 13 | 3217 | 92 | |
XM_017020706.1 | XP_016876195.1 | 9 - 10 | 1336 | 92 | |
GenBank mRNA | AB051508.1 | - | 15 - 16 | 2148 | 92 |
AK023108.1 | - | 7 - 8 | 1141 | 92 | |
AK057924.1 | - | 15 - 16 | 2155 | 92 | |
BC042504.1 | - | 12 - 13 | 1721 | 92 | |
BC050680.1 | - | 12 - 13 | 1738 | 92 | |
BC113571.1 | - | 14 - 15 | 2088 | 92 | |
BC113573.1 | - | 14 - 15 | 2088 | 92 | |
BX647843.1 | - | 6 - 7 | 956 | 92 |