Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNFT2
Entrez Gene ID
84900
Gene Name
ring finger protein, transmembrane 2
Gene Aliases
TMEM118
Chromosome Location
Chr.12: 116738291 - 116853631 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109903.1 NP_001103373.1 4 - 5 788 59
NM_032814.3 NP_116203.2 4 - 5 788 59
GenBank mRNA AK027533.1 - 5 - 6 824 59
BC011878.2 - 4 - 5 768 59
BQ676121.1 - 1 - 2 459 59

View Full Product Details