The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109903.1 | NP_001103373.1 | 8 - 9 | 1267 | 58 |
NM_032814.3 | NP_116203.2 | 8 - 9 | 1267 | 58 | |
GenBank mRNA | AK027533.1 | - | 9 - 10 | 1303 | 58 |
AK093164.1 | - | 4 - 5 | 1068 | 58 |