The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184768.1 | NP_001171697.1 | 1 - 2 | 119 | 69 |
NM_019007.3 | NP_061880.2 | 1 - 2 | 119 | 69 | |
NR_033669.1 | 1 - 2 | 119 | 69 | ||
NR_033670.1 | 1 - 2 | 119 | 69 | ||
XM_011530976.2 | XP_011529278.1 | 1 - 2 | 94 | 69 | |
GenBank mRNA | BC007677.2 | - | 1 - 2 | 119 | 69 |
BX423052.2 | - | 1 - 2 | 120 | 69 | |
DA933518.1 | - | 1 - 2 | 116 | 69 |