The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039547.2 | NP_001034636.1 | 1 - 2 | 297 | 101 |
NR_033289.1 | 1 - 2 | 297 | 101 | ||
XM_017006088.1 | XP_016861577.1 | 1 - 2 | 282 | 101 | |
XM_017006089.1 | XP_016861578.1 | 1 - 2 | 265 | 101 | |
XM_017006090.1 | XP_016861579.1 | 1 - 2 | 282 | 101 | |
GenBank mRNA | AK127641.1 | - | 1 - 2 | 265 | 101 |
AK310761.1 | - | 1 - 2 | 282 | 101 | |
AK313792.1 | - | 1 - 2 | 179 | 101 | |
BC032470.1 | - | 1 - 2 | 257 | 101 | |
BX648359.1 | - | 1 - 2 | 277 | 101 | |
BX648681.1 | - | 1 - 2 | 238 | 101 | |
DR000158.1 | - | 1 - 2 | 298 | 101 |