Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNM1
Entrez Gene ID
1759
Gene Name
dynamin 1
Gene Aliases
DNM, EIEE31
Chromosome Location
Chr.9: 128203355 - 128255248 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288737.1 NP_001275666.1 10 - 11 1451 78
NM_001288738.1 NP_001275667.1 10 - 11 1451 78
NM_001288739.1 NP_001275668.1 10 - 11 1451 78
XM_005251768.2 XP_005251825.1 10 - 11 1451 78
XM_006716992.2 XP_006717055.1 10 - 11 1451 78
XM_006716993.1 XP_006717056.1 10 - 11 1451 78
XM_011518335.2 XP_011516637.1 10 - 11 1451 78
GenBank mRNA AK296326.1 - 10 - 11 1418 78
AK308793.1 - 3 - 4 252 78
AK316235.1 - 10 - 11 1427 78
L07808.1 - 2 - 3 177 78

View Full Product Details