The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052891.2 | NP_443123.1 | 4 - 5 | 583 | 64 |
XM_011509118.1 | XP_011507420.1 | 6 - 7 | 1012 | 64 | |
XM_011509119.1 | XP_011507421.1 | 5 - 6 | 904 | 64 | |
XM_011509120.2 | XP_011507422.1 | 6 - 7 | 790 | 64 | |
GenBank mRNA | AY035376.1 | - | 4 - 5 | 583 | 64 |
BC069741.1 | - | 4 - 5 | 569 | 64 | |
BC069798.1 | - | 4 - 5 | 570 | 64 | |
BC069801.1 | - | 4 - 5 | 570 | 64 | |
BC128114.1 | - | 4 - 5 | 556 | 64 | |
BC128115.1 | - | 4 - 5 | 556 | 64 |