Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPDZ
Entrez Gene ID
8777
Gene Name
multiple PDZ domain crumbs cell polarity complex component
Gene Aliases
HYC2, MUPP1
Chromosome Location
Chr.9: 13105296 - 13279687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261406.1 NP_001248335.1 35 - 36 5077 102
NM_001261407.1 NP_001248336.1 35 - 36 5077 102
NM_003829.4 NP_003820.2 36 - 37 5176 102
XM_005251622.4 XP_005251679.1 36 - 37 5125 102
XM_005251623.4 XP_005251680.1 34 - 35 4915 102
XM_006716885.3 XP_006716948.1 36 - 37 5174 102
XM_006716886.3 XP_006716949.1 37 - 38 5333 102
XM_006716887.3 XP_006716950.1 36 - 37 5277 102
XM_006716888.3 XP_006716951.1 36 - 37 5125 102
XM_006716889.3 XP_006716952.1 35 - 36 5029 102
XM_006716891.3 XP_006716954.1 35 - 36 5029 102
XM_017015253.1 XP_016870742.1 35 - 36 5014 102
XM_017015254.1 XP_016870743.1 35 - 36 5126 102
XM_017015255.1 XP_016870744.1 34 - 35 4974 102
XM_017015256.1 XP_016870745.1 34 - 35 4915 102
XM_017015257.1 XP_016870746.1 34 - 35 4902 102
GenBank mRNA AB210041.1 - 36 - 37 5177 102
AF093419.1 - 35 - 36 5000 102
AJ001319.1 - 1 - 2 103 102
AK091945.1 - 11 - 12 1454 102
AK098775.1 - 3 - 4 354 102
AK294823.1 - 9 - 10 1086 102
AK294976.1 - 14 - 15 1682 102
AK307924.1 - 9 - 10 1085 102
AK307988.1 - 12 - 13 1494 102
AK316489.1 - 13 - 14 1560 102
BC140793.1 - 34 - 35 4901 102
BC144564.1 - 34 - 35 4901 102
CR936648.1 - 20 - 21 5064 102

View Full Product Details