Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPDZ
Entrez Gene ID
8777
Gene Name
multiple PDZ domain crumbs cell polarity complex component
Gene Aliases
HYC2, MUPP1
Chromosome Location
Chr.9: 13105296 - 13279687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261406.1 NP_001248335.1 43 - 44 5952 94
NM_001261407.1 NP_001248336.1 42 - 43 5865 94
NM_003829.4 NP_003820.2 43 - 44 5964 94
XM_005251622.4 XP_005251679.1 44 - 45 6000 94
XM_005251623.4 XP_005251680.1 42 - 43 5790 94
XM_006716885.3 XP_006716948.1 44 - 45 6049 94
XM_006716886.3 XP_006716949.1 45 - 46 6208 94
XM_006716887.3 XP_006716950.1 44 - 45 6152 94
XM_006716888.3 XP_006716951.1 43 - 44 5913 94
XM_006716889.3 XP_006716952.1 43 - 44 5904 94
XM_006716891.3 XP_006716954.1 42 - 43 5817 94
XM_017015252.1 XP_016870741.1 43 - 44 5916 94
XM_017015253.1 XP_016870742.1 42 - 43 5802 94
XM_017015254.1 XP_016870743.1 43 - 44 6001 94
XM_017015255.1 XP_016870744.1 42 - 43 5849 94
XM_017015256.1 XP_016870745.1 41 - 42 5703 94
XM_017015257.1 XP_016870746.1 41 - 42 5690 94
GenBank mRNA AB210041.1 - 43 - 44 5965 94
AF093419.1 - 43 - 44 5791 94
AJ001319.1 - 9 - 10 978 94
AK058011.1 - 4 - 5 1391 94
AK074721.1 - 13 - 14 1453 94
AK091945.1 - 19 - 20 2329 94
AK098775.1 - 11 - 12 1229 94
AK294823.1 - 17 - 18 1961 94
AK294976.1 - 22 - 23 2557 94
AK307988.1 - 20 - 21 2369 94
AK316489.1 - 21 - 22 2435 94
BC140793.1 - 42 - 43 5776 94
BC144564.1 - 41 - 42 5689 94
CR936648.1 - 28 - 29 5939 94

View Full Product Details