The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181900.2 | NP_871629.1 | 4 - 5 | 484 | 64 |
NR_135013.1 | 3 - 4 | 434 | 64 | ||
XM_017022639.1 | XP_016878128.1 | 3 - 4 | 999 | 64 | |
GenBank mRNA | AF480304.1 | - | 4 - 5 | 400 | 64 |
AK026352.1 | - | 3 - 4 | 392 | 64 | |
AK225479.1 | - | 3 - 4 | 392 | 64 | |
AL137657.1 | - | 3 - 4 | 434 | 64 | |
BC004365.1 | - | 4 - 5 | 426 | 64 |