The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028045.1 | 2 - 3 | 480 | 80 | |
GenBank mRNA | AF047445.1 | - | 2 - 3 | 210 | 80 |
AK290077.1 | - | 1 - 2 | 181 | 80 | |
AK296103.1 | - | 2 - 3 | 217 | 80 | |
BC069672.1 | - | 2 - 3 | 183 | 80 | |
BC069697.1 | - | 2 - 3 | 183 | 80 | |
BC069734.1 | - | 2 - 3 | 183 | 80 | |
BC101471.1 | - | 2 - 3 | 197 | 80 | |
BC101473.1 | - | 2 - 3 | 200 | 80 | |
DA776936.1 | - | 2 - 3 | 480 | 80 |